Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

73820

Sigma-Aldrich

5-Nitrosalicylic acid

purum, ≥98.0% (T)

Synonym(s):

Anilotic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H3(NO2)CO2H
CAS Number:
Molecular Weight:
183.12
Beilstein:
2213719
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Assay

≥98.0% (T)

mp

227-233 °C
228-230 °C (lit.)

SMILES string

OC(=O)c1cc(ccc1O)[N+]([O-])=O

InChI

1S/C7H5NO5/c9-6-2-1-4(8(12)13)3-5(6)7(10)11/h1-3,9H,(H,10,11)

InChI key

PPDRLQLKHRZIJC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

This product has been replaced by 247871-ALDRICH | 2-Hydroxy-5-nitrobenzoic acid 99%

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

73820-INTR:
73820-25G:
73820-100G:
73820-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service