Skip to Content
Merck
All Photos(1)

Key Documents

73617

Sigma-Aldrich

4-Nitrophenyl β-D-cellobioside

Synonym(s):

p-Nitrophenyl β-D-cellobioside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H25NO13
CAS Number:
Molecular Weight:
463.39
Beilstein:
100234
MDL number:
UNSPSC Code:
12352204

optical activity

[α]20/D −86±2°, c = 1% in methanol: water (2:3)

impurities

≤0.05% free 4-nitrophenol

solubility

methanol: water (2:3): 50 mg/mL, clear, faintly yellow

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)[C@@H](O[C@@H]2CO)Oc3ccc(cc3)[N+]([O-])=O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C18H25NO13/c20-5-9-11(22)12(23)14(25)18(30-9)32-16-10(6-21)31-17(15(26)13(16)24)29-8-3-1-7(2-4-8)19(27)28/h1-4,9-18,20-26H,5-6H2/t9-,10-,11-,12+,13-,14-,15-,16-,17-,18+/m1/s1

InChI key

IAYJZWFYUSNIPN-KFRZSCGFSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Substrate for the measurement of exoglucanase activity

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M V Deshpande et al.
Analytical biochemistry, 138(2), 481-487 (1984-05-01)
A selective procedure using synthetic substrates for determination of exo-1,4,-beta-glucanases in a mixture of exoglucanases , endoglucanases , and beta-glucosidases is formulated. The heterobiosides , p- nithrophenyl -beta-D- cellobioside ( pNPC ) or p-nitrophenyl-beta-D-lactoside ( pNPL ), were used as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service