Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

73307

Supelco

2-Nitrophenyl phenyl ether

Selectophore, ≥99.0%

Synonym(s):

2-Nitrodiphenyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H9NO3
CAS Number:
Molecular Weight:
215.20
Beilstein:
2051914
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

product line

Selectophore

Quality Level

Assay

≥99.0% (GC)
≥99.0%

refractive index

n20/D 1.600

bp

106-108 °C/0.01 mmHg (lit.)

density

1.241 g/mL at 20 °C (lit.)

SMILES string

[O-][N+](=O)c1ccccc1Oc2ccccc2

InChI

1S/C12H9NO3/c14-13(15)11-8-4-5-9-12(11)16-10-6-2-1-3-7-10/h1-9H

InChI key

VNHGETRQQSYUGZ-UHFFFAOYSA-N

General description

Visit our Sensor Applications portal to learn more.

Other Notes

Plasticizer in polymer membrane neutral carrier-based ion-selective electrodes

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 4: Flammable liquids
Type 3 petroleums
Hazardous rank III
Water insoluble liquid

JAN Code

73307-5ML:
73307-VAR:
73307-BULK:
73307-1ML:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T. Katsu et al.
Analytica Chimica Acta, 217, 193-193 (1989)
Lithium-selective polymeric membrane electrodes based on dodecylmethyl-14-crown-4.
S Kitazawa et al.
The Analyst, 110(3), 295-299 (1985-03-01)
Guanidinium ion-selective electrodes based on dibenzo-27-crown-9 and tetraphenylborate.
F N Assubaie et al.
The Analyst, 113(1), 61-64 (1988-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service