Skip to Content
Merck
All Photos(1)

Key Documents

61820

Sigma-Aldrich

L-Leucine

≥99.0% (NT)

Synonym(s):

(S)-2-Amino-4-methylpentanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
131.17
Beilstein:
1721722
EC Number:
MDL number:
UNSPSC Code:
12352209

Assay

≥99.0% (NT)

optical activity

[α]20/D +15.5±1.0°, c = 5% in 5 M HCl

impurities

≤0.3% foreign amino acids

ign. residue

≤0.1% (as SO4)

loss

≤0.1% loss on drying, 110 °C

mp

>300 °C (lit.)

anion traces

chloride (Cl-): ≤200 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
NH4+: ≤100 mg/kg
Na: ≤100 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

CC(C)C[C@H](N)C(O)=O

InChI

1S/C6H13NO2/c1-4(2)3-5(7)6(8)9/h4-5H,3,7H2,1-2H3,(H,8,9)/t5-/m0/s1

InChI key

ROHFNLRQFUQHCH-YFKPBYRVSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marion Lanier et al.
Bioorganic & medicinal chemistry, 18(2), 573-579 (2009-12-25)
Placental Alkaline Phosphatase (PLAP) is a tissue-restricted isozyme of the Alkaline Phosphatase (AP) superfamily. PLAP is an oncodevelopmental enzyme expressed during pregnancy and in a variety of human cancers, but its biological function remains unknown. We report here a series
Alex Avdeef et al.
Journal of medicinal chemistry, 53(9), 3566-3584 (2010-04-09)
The study aimed to predict effective human jejunal permeability (P(eff)) using a biophysical model based on parametrized paracellular, aqueous boundary layer, and transcellular permeabilities, and the villus-fold surface area expansion factor (k(VF)). Published human jejunal data (119 P(eff), 53 compounds)
Iris Thondorf et al.
Bioorganic & medicinal chemistry, 19(21), 6409-6418 (2011-10-01)
The proton-coupled amino acid transporter hPAT1 has recently gained much interest due to its ability to transport small drugs thereby allowing their oral administration. A three-dimensional quantitative structure-activity relationship (3D QSAR) study has been performed on its natural and synthetic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service