Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

56619

Supelco

Bacopaside I

analytical standard

Synonym(s):

Pseudojujubogenin 3-O-[α-L-arabinofuranosyl-(1→2)-[6-O-sulfo-β-D-glucopyranosyl-(1→3)]-α-L-arabinopyranoside]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H74O20S
CAS Number:
Molecular Weight:
979.13
UNSPSC Code:
85151701
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

InChI

1S/C46H74O20S/c1-21(2)14-22-16-59-46-19-45(20-60-46)23(37(46)44(22,7)54)8-9-28-42(5)12-11-29(41(3,4)27(42)10-13-43(28,45)6)64-40-36(66-38-33(52)30(49)25(15-47)62-38)35(24(48)17-58-40)65-39-34(53)32(51)31(50)26(63-39)18-61-67(55,56)57/h14,22-40,47-54H,8-13,15-20H2,1-7H3,(H,55,56,57)/t22-,23-,24+,25+,26-,27+,28-,29+,30+,31-,32+,33-,34-,35+,36-,37+,38+,39+,40+,42+,43-,44+,45+,46?/m1/s1

InChI key

SKFWOYHZBNAJGA-VLNRPALJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bacopaside I belongs to the group of saponins, isolated from Bacopa monnieri plant widely used in promoting mental health, and relief to patients with anxiety or epileptic disorders.

Application

Bacopaside I may be used as an analytical reference standard for the quantification of the analyte in Bacopa monnieri plant extract using high-performance liquid chromatography technique.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

56619-VAR:
56619-BULK:
56619-5MG:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stability studies of crude plant material of Bacopa monnieri and quantitative determination of bacopaside I and bacoside A by HPLC
Srivastava P, et al.
Phytochemical Analysis, 23(5), 502-507 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service