Skip to Content
Merck
All Photos(1)

Documents

Safety Information

52383

Supelco

[(3R)-3-Hydroxy-hexadec-9-enoyl]-L-carnitine

analytical standard

Synonym(s):

(2R)-3-Carboxy-2-[[3R-3-hydroxy-1-oxo-9-hexadecen-1-yl]oxy]-N,N,N-trimethyl-1-propanaminium inner salt, (3R)-3-Hydroxypalmitoleyl-L-carnitine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H43NO5
Molecular Weight:
413.59
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Assay

≥85.0% (HPLC)

form

solid

optical activity

[α]/D -20±2°, c = 0.1 in methanol

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing

storage temp.

−20°C

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

52383-50MG:
52383-1MG:
52383-BULK:
52383-VAR:
52383-10MG:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service