Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

51807

Supelco

Alkannin

analytical standard

Synonym(s):

(−)-2-(1-Hydroxy-4-methyl-3-pentenyl)-5,8-dihydroxy-1,4-naphthoquinone, 5,8-Dihydroxy-2-[(1S)-1-hydroxy-4-methyl-3-penten-1-yl]-1,4-naphthalenedione, Anchusin, C.I. Natural Red 20, E103

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16O5
CAS Number:
Molecular Weight:
288.30
Colour Index Number:
75530
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

grade

analytical standard

Assay

≥95.0% (HPLC)

optical purity

enantiomeric excess: ≥98.0%

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤1.0% water (calc. from elemental analysis)

application(s)

cleaning products
cosmetics
food and beverages
personal care

SMILES string

O=C1C([C@H](CC=C(C)C)O)=CC(C2=C1C(O)=CC=C2O)=O

InChI

1S/C16H16O5/c1-8(2)3-4-10(17)9-7-13(20)14-11(18)5-6-12(19)15(14)16(9)21/h3,5-7,10,17-19H,4H2,1-2H3/t10-/m0/s1

InChI key

NEZONWMXZKDMKF-JTQLQIEISA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

51807-10MG:
51807-BULK:
51807-VAR:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoshitaka Hosokawa et al.
Cell biology international, 40(12), 1380-1385 (2016-10-25)
Alkannin, which is found in Alkanna tinctoria, a member of the borage family, is used as a food coloring. Alkannin has recently been reported to have certain biological functions, such as anti-microbial and anti-oxidant effects. It is known that CC
Cheng-Yi Tang et al.
Molecular ecology resources, 20(1), 228-241 (2019-10-19)
Boraginales (the forget-me-not order) is a core group within the lamiids clade. However, until now, no genome from Boraginales has been reported, and published transcriptomes are also rare. Here, we report the first Boraginales species de novo genome (i.e. Echium
Yanbin Liu et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 37(10), 13649-13657 (2016-07-31)
Von Hippel-Lindau (VHL) is the most frequently mutated gene in clear cell renal carcinoma. Here, we identified a novel translational variant of VHL, termed VHLα, initiated from an alternative translational start site upstream and in frame with the ATG start
Xiao-Fei Shang et al.
Scientific reports, 8(1), 1609-1609 (2018-01-27)
As important secondary plant metabolites, naphthoquinones exhibit a wide range of biological activities. However, their potential as sustainable alternatives to synthetic acaricides has not been studied. This study for the first time investigates the acaricidal activity of naphthoquinones against Psoroptes
Anshul Kundaje et al.
Nature, 518(7539), 317-330 (2015-02-20)
The reference human genome sequence set the stage for studies of genetic variation and its association with human disease, but epigenomic studies lack a similar reference. To address this need, the NIH Roadmap Epigenomics Consortium generated the largest collection so

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service