Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

51295

Millipore

Haemophilus Medium

suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171607

form

powder

Quality Level

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing
food and beverages

microbiology

suitability

Haemophilus spp.

Application

Optimized medium for the growth of Haemophilus species.

Preparation Note

Supplements :
44 ml/l glycerol (Cat. No. 49767)
10 mg/l β-NAD (Cat. No. 43410)
30 mg/l hemin (Cat. No. 51280)

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

51295-BULK:
51295-100G:
51295-500G:
51295-VAR:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service