Skip to Content
Merck
All Photos(1)

Key Documents

50110

Sigma-Aldrich

Glycine methyl ester hydrochloride

puriss., ≥99.0% (AT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2COOCH3 · HCl
CAS Number:
Molecular Weight:
125.55
Beilstein:
3593644
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

puriss.

Assay

≥99.0% (AT)

ign. residue

≤0.05%

mp

175 °C (dec.) (lit.)
175-176 °C (dec.)

SMILES string

Cl.COC(=O)CN

InChI

1S/C3H7NO2.ClH/c1-6-3(5)2-4;/h2,4H2,1H3;1H

InChI key

COQRGFWWJBEXRC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Iwama et al.
Biological & pharmaceutical bulletin, 24(9), 978-981 (2001-09-18)
The sialic acid binding lectin from bullfrog Rana catesbeiana oocyte (cSBL) is known to have anti-tumor activity. In order to investigate the relationship between the net charge of cSBL and its anti-tumor effect, cSBL was modified with a water-soluble carbodiimide
Wenzhong Gao et al.
Organic letters, 7(19), 4241-4244 (2005-09-09)
[reaction: see text] A novel Cu(I)-catalyzed asymmetric 1,3-dipolar cycloaddition of azomethine ylides with acrylates has been developed. Up to 98/2 exo/endo selectivity and up to 98% enantiomeric excess have been achieved.
Akihiko Tohri et al.
European journal of biochemistry, 271(5), 962-971 (2004-03-11)
To elucidate the domains on the extrinsic 23 kDa protein involved in electrostatic interaction with the extrinsic 33 kDa protein in spinach photosystem II, we modified amino or carboxyl groups of the 23 kDa protein to uncharged methyl ester groups
J M Delfino et al.
International journal of peptide and protein research, 21(4), 440-450 (1983-04-01)
The reactivity of the carboxyl groups in bovine growth hormone was studied by reaction with 1-ethyl-3(3-dimethylaminopropyl) carbodiimide in the presence of an excess of glycinemethylester. Localization in the molecule of the various kinetically distinguishable carboxyl groups was achieved. Highest reactivity
Vincenzo Santagada et al.
Journal of combinatorial chemistry, 7(4), 618-621 (2005-07-12)
An easy and convenient microwave-assisted synthesis of N-alkylated glycine methyl esters is described. Parallel and nonparallel combinatorial methods are described and compared. The described reactions are reductive alkylations of several aldehydes and glycine methyl ester in the presence of NaBH3CN.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service