Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

49954

Supelco

3-Methyl-13C-glutaric acid-1,2,3-13C3

analytical standard

Synonym(s):

3-Methylglutaric acid-13C4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C4C2H10O4
Molecular Weight:
150.11
UNSPSC Code:
41116107

grade

analytical standard

isotopic purity

≥99.0%

Assay

≥95.0% (GC)

form

solid

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing

SMILES string

OC(C[13CH]([13CH3])[13CH2][13C](O)=O)=O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

49954-VAR:
49954-BULK:
49954-5MG:
49954-1MG:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service