Skip to Content
Merck
All Photos(1)

Key Documents

443379

Sigma-Aldrich

Xylenes

ACS reagent, ≥98.5% xylenes + ethylbenzene basis

Synonym(s):

Xylene mixture of isomers

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CH3)2
CAS Number:
Molecular Weight:
106.17
EC Number:
MDL number:
UNSPSC Code:
12190000

grade

ACS reagent

vapor density

3.7 (vs air)

vapor pressure

18 mmHg ( 37.7 °C)

Assay

≥98.5% xylenes + ethylbenzene basis

autoignition temp.

867 °F

expl. lim.

7 %

impurities

H2SO4, passes test (substances darkened by sulfuric acid)
≤0.003% S compounds
≤0.05% water
≤25% C6H5CH2CH3

evapn. residue

≤0.002%

color

APHA: ≤10

refractive index

n20/D 1.497 (lit.)

bp

137-140 °C (lit.)

density

0.86 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(C)cc1.Cc2cccc(C)c2.Cc3ccccc3C

InChI

1S/3C8H10/c1-7-3-5-8(2)6-4-7;1-7-4-3-5-8(2)6-7;1-7-5-3-4-6-8(7)2/h3*3-6H,1-2H3

InChI key

MVZVDAGWAAZJPE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anne-France le Rolle et al.
Journal of translational medicine, 13, 199-199 (2015-06-25)
Clinical therapy for metastatic colorectal cancer (CRC) remains limited, especially when the tumor harbors a KRAS mutation. This study aimed to identify prognostic biomarkers in CRC that are accessible for therapeutic inhibition. Conditioned media from human CRC epithelial cells and
Ryan P Smith et al.
Human gene therapy, 26(6), 377-385 (2015-03-27)
Much research has gone into the development of hybrid gene delivery systems that combine the broad tropism and efficient transduction of adenoviral vectors with the ability to achieve stable expression of cargo genes. In addition to gene therapy applications, such
Osceola Whitney et al.
Developmental neurobiology, 75(7), 778-790 (2014-11-20)
The forkhead domain FOXP2 and FOXP1 transcription factors are implicated in several cognitive disorders with language deficits, notably autism, and thus play a central role in learned vocal motor behavior in humans. Although a similar role for FoxP2 and FoxP1
Chani M Becker et al.
Neuro-oncology, 17(9), 1210-1219 (2015-05-15)
Targeting drug delivery to invasive glioma cells is a particularly difficult challenge because these cells lie behind an intact blood-brain barrier (BBB) that can be observed using multimodality imaging. BBB-associated efflux transporters such as P-glycoprotein (P-gp) and breast cancer resistance
Qiming Wang et al.
Nature communications, 5, 4899-4899 (2014-09-17)
Cephalopods can display dazzling patterns of colours by selectively contracting muscles to reversibly activate chromatophores--pigment-containing cells under their skins. Inspired by this novel colouring strategy found in nature, we design an electro-mechano-chemically responsive elastomer system that can exhibit a wide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service