Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

42696

Supelco

Sieve Standard, traceable to NIST SRM

75 μm (200 mesh)

Synonym(s):

Sieve calibration standard, 75 μm (200 mesh)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

form

particles

manufacturer/tradename

Whitehouse Scientific Ltd SS398

particle size

75 μm (200 mesh)

application(s)

glass & ceramic
industrial qc
pharmaceutical

format

neat

Looking for similar products? Visit Product Comparison Guide

General description

Sieve Standard, traceable to NIST SRM is designed to find the particle size distribution (PSD) of a particle system.
The particle diameter of 75 μm (200 mesh) is evaluated using electroformed sieves and glass microsphere method.

Application

Used to develop and standardize surface scanning instruments and particle counters.
Also suitable for use:
  • as a sieve calibration standard in electroformed sieve particle size measurements[1]
  • to sieve aluminum buffing residue in studies of dust explosion hazards[2]

Features and Benefits

  • suitable for routine instrument calibration checks, testing and corrections
  • certified against NIST and SRM standards
  • available as a neat sample

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

42696-5X1G:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reducing aluminum dust explosion hazards: case study of dust inerting in an aluminum buffing operation
Myers TJ
Journal of Hazardous Materials, 159(1), 72-80 (2008)
Sieve Calibration-a New Simple but High Precision Approach
Rideal GR, et al.
Particle & Particle Systems Characterization, 17(2), 77-82 (2000)

Related Content

Particle size conversion table from our Lab Basics Technical Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service