Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

41368

Supelco

p-Toluenesulfonyl isocyanate

for HPLC derivatization, LiChropur, ≥98.0% (GC)

Synonym(s):

p-Tosyl isocyanate, TsNCO, Tosyl isocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4SO2NCO
CAS Number:
Molecular Weight:
197.21
Beilstein:
391287
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.22

grade

for HPLC derivatization

Quality Level

vapor pressure

1 mmHg ( 100 °C)

Assay

≥98.0% (GC)

form

liquid

quality

LiChropur

technique(s)

HPLC: suitable

refractive index

n20/D 1.533-1.537
n20/D 1.534 (lit.)

bp

144 °C/10 mmHg (lit.)
275-278 °C (lit.)

density

1.291 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(cc1)S(=O)(=O)N=C=O

InChI

1S/C8H7NO3S/c1-7-2-4-8(5-3-7)13(11,12)9-6-10/h2-5H,1H3

InChI key

VLJQDHDVZJXNQL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

p-Toluenesulfonyl isocyanate, a sulfonyl isocyanate derivative, is a highly reactive compound that finds application as a water scavenger. It is used as a component of polyurethane coatings, sealants, adhesives and as an intermediate of several industrially important chemicals.

Application

p-Toluenesulfonyl isocyanate may be used as a derivatization reagent for the analyses of diethylene glycol and propylene glycol in pharmaceutical products, stereoisomers of 3-hydroxyl-7-methyl-norethynodrel in biological samples, and pyrrolizidine alkaloids in herbal tea formulations by high performance liquid chromatography (HPLC) based techniques.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

294.8 °F - closed cup

Flash Point(C)

146 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 4: Flammable liquids
Type 3 petroleums
Hazardous rank III
Water insoluble liquid

JAN Code

41368-1ML:
41368-BULK:
41368-VAR:
41368-10ML:
41368-1ML-KC:
41368-10X1ML:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Parimala et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 81(1), 711-723 (2011-07-29)
The Fourier transform infrared (FTIR) and FT Raman spectra of p-toluenesulfonyl isocyanate (p-tosyl isocyanate) have been measured. The molecular geometry, vibrational frequencies, infrared intensities, Raman activities and atomic charges have been calculated by using ab initio HF and density functional
Inga Mädge et al.
Food chemistry, 187, 491-498 (2015-05-16)
A general contamination of tea with pyrrolizidine alkaloids (PA) has just become known. Here, we report the application and modification of a new HPLC-ESI-MS/MS sum parameter method to quantitate PA content of herbal teas intended for infants, pregnant and lactating
Ming Zuo et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 814(2), 331-337 (2005-01-11)
In this paper, p-toluenesulfonyl isocyanate has been reported as a novel derivatization reagent with strong nuclephilic reactivity for the hydroxyl compounds. The derivatization for the two pharmacologically active 3-hydroxyl metabolites, 3alpha-hydroxyl-7-methyl-norethynodrel and 3beta-hydroxyl-7-methyl-norethynodrel by p-toluenesulfonyl isocyanate can be accomplished in
Tao Zhou et al.
Journal of separation science, 30(16), 2620-2627 (2007-09-21)
A simple and reliable HPLC method was developed for the simultaneous quantitative analysis of diethylene glycol (DEG) and propylene glycol (PG) in pharmaceutical products by precolumn derivatization. The derivatization reagent p-toluenesulfonyl isocyanate (TSIC, 10 microL, 20% in ACN v/v) was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service