34567
Flonicamid Metabolite TFNA
PESTANAL®, analytical standard
Synonym(s):
4-(Trifluoromethyl)pyridine-3-carboxylic acid, 4-(Trifluoromethyl)nicotinic acid
About This Item
Recommended Products
grade
analytical standard
product line
PESTANAL®
mp
146-148 °C (lit.)
application(s)
agriculture
environmental
SMILES string
OC(=O)c1cnccc1C(F)(F)F
InChI
1S/C7H4F3NO2/c8-7(9,10)5-1-2-11-3-4(5)6(12)13/h1-3H,(H,12,13)
InChI key
LMRJHNFECNKDKH-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
JAN Code
34567-VAR:
34567-BULK:
34567-25MG:
34567-BULK-MG:
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service