34109
Meloxicam-d3
VETRANAL®, analytical standard
Synonym(s):
4-Hydroxy-2-(methyl-d3)-1,1-dioxo-benzo[e]-1,2-thiazine-3-carboxylic acid (5-methyl-2-thiazolyl)amide
About This Item
Recommended Products
grade
analytical standard
Quality Level
product line
VETRANAL®
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
clinical testing
format
neat
mass shift
M+3
SMILES string
[2H]C([2H])([2H])N1C(C(=O)Nc2ncc(C)s2)=C(O)c3ccccc3S1(=O)=O
InChI
1S/C14H13N3O4S2/c1-8-7-15-14(22-8)16-13(19)11-12(18)9-5-3-4-6-10(9)23(20,21)17(11)2/h3-7,18H,1-2H3,(H,15,16,19)/i2D3
InChI key
ZRVUJXDFFKFLMG-BMSJAHLVSA-N
Application
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 2
Personal Protective Equipment
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
JAN Code
34109-50MG:
34109-VAR:
34109-BULK:
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service