Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

32637

Supelco

S-Metolachlor Metabolite CGA 357704

PESTANAL®, analytical standard

Synonym(s):

(S)-2-[(Oxalyl)(2-ethyl-6-methylphenyl)amino]propionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H17NO5
CAS Number:
Molecular Weight:
279.29
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCc1cccc(C)c1N([C@@H](C)C(O)=O)C(=O)C(O)=O

InChI

1S/C14H17NO5/c1-4-10-7-5-6-8(2)11(10)15(9(3)13(17)18)12(16)14(19)20/h5-7,9H,4H2,1-3H3,(H,17,18)(H,19,20)/t9-/m0/s1

InChI key

IMFSUYMDPTXKCC-VIFPVBQESA-N

Application

Dimethachlor Metabolite CGA 373464 can be used as an analytical reference standard for the analysis of the analyte in surface water and groundwater using direct injection liquid chromatography-electrospray ionization-mass spectrometry (LC-ESI-MS) under multiple reaction monitoring (MRM) mode.[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

32637-BULK:
32637-VAR:
32637-100MG:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A multimethod for the determination of 150 pesticide metabolites in surface water and groundwater using direct injection liquid chromatography-mass spectrometry.
Reemtsma, et al.
Journal of Chromatography A, 1271(1), 95-104 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service