Skip to Content
Merck
All Photos(1)

Key Documents

32442

Supelco

Amitraz Metabolite BTS 27271

PESTANAL®, analytical standard

Synonym(s):

N-(2,4-Dimethylphenyl)-N′-methylformamidine, N-Methyl-N′-2,4-xylylformamidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N2
CAS Number:
Molecular Weight:
162.23
Beilstein:
1210087
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

product line

PESTANAL®

application(s)

agriculture
environmental

SMILES string

CN\C=N\c1ccc(C)cc1C

InChI

1S/C10H14N2.ClH/c1-8-4-5-10(9(2)6-8)12-7-11-3;/h4-7H,1-3H3,(H,11,12);1H

InChI key

VXSNJXDZTGFDMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E A Abu-Basha et al.
Metabolism: clinical and experimental, 48(11), 1461-1469 (1999-12-03)
The study purpose was to investigate the direct effect of amitraz, a formamidine insecticide/acaricide, and its active metabolite BTS 27271 on insulin and glucagon secretion from the perfused rat pancreas. Amitraz and BTS 27271 (0.01, 0.1, 1, and 10 micromol/L)
M A Pass et al.
Comparative biochemistry and physiology. C, Comparative pharmacology and toxicology, 99(1-2), 169-172 (1991-01-01)
1. Amitraz was rapidly metabolised to BTS27271 after intravenous administration to sheep. 2. Amitraz and BTS27271 had significant H1-histamine antagonist activity on isolated guinea-pig ileum. BTS27271 was approximately 3.3 times as potent as amitraz. 3. Intravenous injection of amitraz and
Metabolic conversion of N'-(2,4-dimethylphenyl)-N-methylformamidine pesticide and the analysis of the metabolites.
Z Naikai et al.
Bulletin of environmental contamination and toxicology, 65(1), 22-27 (2000-06-30)
Carmen Ferrer et al.
Journal of AOAC International, 93(2), 380-388 (2010-05-20)
This paper compares the performance of the three most widely employed multiresidue methods [quick, easy, cheap, effective, rugged, and safe (QuEChERS), mini-Luke, and ethyl acetate] currently used for the determination of amitraz residues in fruits. A fast and differentiated analysis
A F Lehner et al.
Journal of analytical toxicology, 28(7), 553-562 (2004-11-02)
Amitraz (N'-(2,4-dimethylphenyl)-N-[[(2,4-dimethylphenyl)imino]methyl]-N-methyl-methanimidamide) is an alpha-2 adrenergic agonist used in veterinary medicine primarily as a scabicide- or acaricide-type insecticide. As an alpha-2 adrenergic agonist, it also has sedative/tranquilizing properties and is, therefore, listed as an Association of Racing Commissioners International Class

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service