32316
Magnesium acetate tetrahydrate
puriss. p.a., ACS reagent, 99.5-102%
Synonym(s):
Acetic acid magnesium salt
About This Item
Recommended Products
grade
ACS reagent
puriss. p.a.
Assay
99.5-102%
form
solid
impurities
≤0.0005% heavy metals (as Pb)
≤0.005% insoluble in acetic acid
pH
8-9 (20 °C, 5%)
mp
72-75 °C (lit.)
anion traces
chloride (Cl-): ≤10 mg/kg
sulfate (SO42-): ≤50 mg/kg
cation traces
Ba: ≤10 mg/kg
Ca: ≤100 mg/kg
Cd: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤10 mg/kg
Mn: ≤10 mg/kg
Na: ≤10 mg/kg
Pb: ≤5 mg/kg
Sr: ≤10 mg/kg
Zn: ≤5 mg/kg
SMILES string
O.O.O.O.CC(=O)O[Mg]OC(C)=O
InChI
1S/2C2H4O2.Mg.4H2O/c2*1-2(3)4;;;;;/h2*1H3,(H,3,4);;4*1H2/q;;+2;;;;/p-2
InChI key
XKPKPGCRSHFTKM-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
General description
Application
Other Notes
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
JAN Code
32316-100G:
32316-250G:
32316-1KG-D:
32316-1.5KG:
32316-6X1KG:
32316-5KG:
32316-2.5KG:
32316-6X100G:
32316-50KG-H-D:
32316-50KG-H:
32316-250G-D:
32316-6X250G:
32316-BULK:
32316-1KG:
32316-25KG-R:
32316-4X5KG:
32316-100G-D:
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service