Skip to Content
Merck
All Photos(1)

Key Documents

31603

Sigma-Aldrich

2,2-Diethoxyacetamide

purum, ≥97.0% (GC)

Synonym(s):

Glyoxylic amide diethyl acetal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2O)2CHCONH2
CAS Number:
Molecular Weight:
147.17
Beilstein:
1760414
EC Number:
MDL number:
UNSPSC Code:
12352100

grade

purum

Assay

≥97.0% (GC)

mp

76-78 °C (lit.)
78-80 °C

SMILES string

CCOC(OCC)C(N)=O

InChI

1S/C6H13NO3/c1-3-9-6(5(7)8)10-4-2/h6H,3-4H2,1-2H3,(H2,7,8)

InChI key

DSKGWFQMJHBHRT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Building block for heterocyclic compounds[1][2]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K. Inami et al.
Bulletin of the Chemical Society of Japan, 58, 352-352 (1985)
J.B. Press et al.
Journal of Heterocyclic Chemistry, 25, 1571-1571 (1988)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service