Skip to Content
Merck
All Photos(1)

Documents

Safety Information

307122

Sigma-Aldrich

4-Amino-3,6-disulfo-1,8-naphthalic anhydride dipotassium salt

Synonym(s):

Lucifer Yellow anhydride dipotassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H5K2NO9S2
CAS Number:
Molecular Weight:
449.50
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder

Quality Level

mp

>300 °C (lit.)

ε (extinction coefficient)

≥20000 at 264-268 nm in 0.1 M NaOH at 0.01 g/L
≥30000 at 226-232 nm in 0.1 M NaOH at 0.01 g/L
≥8000 at 348-352 nm in 0.1 M NaOH at 0.01 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[K+].[K+].Nc1c(cc2C(=O)OC(=O)c3cc(cc1c23)S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C12H7NO9S2.2K/c13-10-5-1-4(23(16,17)18)2-6-9(5)7(12(15)22-11(6)14)3-8(10)24(19,20)21;;/h1-3H,13H2,(H,16,17,18)(H,19,20,21);;/q;2*+1/p-2

InChI key

MIOVHACHHDENNE-UHFFFAOYSA-L

Related Categories

Application

4-Amino-3,6-disulfo-1,8-naphthalic anhydride dipotassium salt has been used to synthesize the fluorescent peptide analog, Lucifer yellow anhydride.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

307122-BULK:
307122-VAR:
307122-100MG:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service