Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

240745

Sigma-Aldrich

Fumaric acid

≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCCH=CHCOOH
CAS Number:
Molecular Weight:
116.07
Beilstein:
605763
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39021709
PubChem Substance ID:

vapor pressure

1.7 mmHg ( 165 °C)

Assay

≥99%

autoignition temp.

1364 °F

expl. lim.

40 %

mp

298-300 °C (subl.) (lit.)

SMILES string

OC(=O)\C=C\C(O)=O

InChI

1S/C4H4O4/c5-3(6)1-2-4(7)8/h1-2H,(H,5,6)(H,7,8)/b2-1+

InChI key

VZCYOOQTPOCHFL-OWOJBTEDSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

523.4 °F

Flash Point(C)

273 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

240745-5G:
240745-100G:
240745-BULK:
240745-1KG:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Comparison of dilute mineral and organic acid pretreatment for enzymatic hydrolysis of wheat straw.
Kootstra AMJ, et al.
Biochemical Engineering Journal, 46(2), 126-131 (2009)
Vibrational spectra and second harmonic generation in molecular complexes of L-lysine with L-tartaric, D, L-malic, acetic, arsenous, and fumaric acids.
Marchewka MK, et al.
Crystal Growth & Design, 3(4), 587-592 (2003)
N Cao et al.
Applied and environmental microbiology, 62(8), 2926-2931 (1996-08-01)
An integrated system of simultaneous fermentation-adsorption for the production and recovery of fumaric acid from glucose by Rhizopus oryzae was investigated. The system was constructed such that growing Rhizopus mycelia were self-immobilized on the plastic discs of a rotary biofilm
Bas J Meussen et al.
Applied microbiology and biotechnology, 94(4), 875-886 (2012-04-25)
Rhizopus oryzae is a filamentous fungus belonging to the Zygomycetes. It is among others known for its ability to produce the sustainable platform chemicals L: -(+)-lactic acid, fumaric acid, and ethanol. During glycolysis, all fermentable carbon sources are metabolized to
Tahar Ghnaya et al.
Chemosphere, 90(4), 1449-1454 (2012-10-03)
The implication of organic acids in Pb translocation was studied in two species varying in shoot lead accumulation, Sesuvium portulacastrum and Brassica juncea. Citric, fumaric, malic and α-cetoglutaric acids were separated and determined by HPLC technique in shoots, roots and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service