22030
L-Carnosine
≥99.0% (NT)
Synonym(s):
β-Alanyl-L-histidine
About This Item
Recommended Products
Assay
≥99.0% (NT)
optical activity
[α]20/D +21±1°, c = 1.5% in H2O
mp
253 °C (dec.) (lit.)
258-260 °C (dec.)
storage temp.
2-8°C
SMILES string
NCCC(=O)N[C@@H](Cc1c[nH]cn1)C(O)=O
InChI
1S/C9H14N4O3/c10-2-1-8(14)13-7(9(15)16)3-6-4-11-5-12-6/h4-5,7H,1-3,10H2,(H,11,12)(H,13,14)(H,15,16)/t7-/m0/s1
InChI key
CQOVPNPJLQNMDC-ZETCQYMHSA-N
Looking for similar products? Visit Product Comparison Guide
Amino Acid Sequence
Biochem/physiol Actions
Other Notes
replaced by
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service