Skip to Content
Merck
All Photos(1)

Key Documents

19724

Sigma-Aldrich

4-tert-Butylcalix[8]arene

purum, ≥95.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C88H112O8
CAS Number:
Molecular Weight:
1297.83
Beilstein:
2612052
MDL number:
UNSPSC Code:
12350000
PubChem Substance ID:

grade

purum

Assay

≥95.0% (HPLC)

mp

411-412 °C (lit.)

SMILES string

CC(C)(C)c1cc2Cc3cc(cc(Cc4cc(cc(Cc5cc(cc(Cc6cc(cc(Cc7cc(cc(Cc8cc(cc(Cc9cc(cc(Cc(c1)c2O)c9O)C(C)(C)C)c8O)C(C)(C)C)c7O)C(C)(C)C)c6O)C(C)(C)C)c5O)C(C)(C)C)c4O)C(C)(C)C)c3O)C(C)(C)C

InChI

1S/C88H112O8/c1-81(2,3)65-33-49-25-51-35-66(82(4,5)6)37-53(74(51)90)27-55-39-68(84(10,11)12)41-57(76(55)92)29-59-43-70(86(16,17)18)45-61(78(59)94)31-63-47-72(88(22,23)24)48-64(80(63)96)32-62-46-71(87(19,20)21)44-60(79(62)95)30-58-42-69(85(13,14)15)40-56(77(58)93)28-54-38-67(83(7,8)9)36-52(75(54)91)26-50(34-65)73(49)89/h33-48,89-96H,25-32H2,1-24H3

InChI key

OLZFZIXORGGLLS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Calixarenes are basket-shaped compounds of potential interest for host-guest complexation studies; Reviews[1]

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C.D. Gutsche
Calixarenes, Monographs in Supramolecular Chemistry (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service