Skip to Content
Merck
All Photos(1)

Documents

Safety Information

18236

Supelco

Ametoctradin hydrochloride Metabolite M650F01

reference material

Synonym(s):

4-(7-Amino-5-ethyl-[1,2,4]triazolo[1,5-a]pyrimidin-6-yl)butanoic acid hydrochloride, Propyl [3-(dimethylamino)propyl]carbamate, Propyl hydrogen [3-(dimethylamino)propyl]carbonimidate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15N5O2 · HCl
CAS Number:
Molecular Weight:
285.73
UNSPSC Code:
41116107

grade

reference material

Quality Level

Assay

≥92.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

format

neat

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

This reference material (RM) is produced and certified in accordance with ISO/IEC 17025. This RM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

18236-10MG:
18236-VAR:
18236-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service