Skip to Content
Merck
All Photos(1)

Key Documents

13904

Sigma-Aldrich

Strontium carbonate

≥96%, white

Synonym(s):

Strontium(II) carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SrCO3
CAS Number:
Molecular Weight:
147.63
EC Number:
MDL number:
UNSPSC Code:
12352302

Assay

≥96%

color

white

density

3.7 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤100 mg/kg
sulfate (SO42-): ≤3000 mg/kg

cation traces

Ba: ≤1.5%
Ca: ≤2000 mg/kg
Fe: ≤50 mg/kg

SMILES string

[Sr++].[O-]C([O-])=O

InChI

1S/CH2O3.Sr/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2

InChI key

LEDMRZGFZIAGGB-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service