Skip to Content
Merck
All Photos(1)

Key Documents

09446

Sigma-Aldrich

Manganese(III) acetylacetonate

technical, ≥95% (KT)

Synonym(s):

2,4-Pentanedione manganese(III) derivative, Manganic acetylacetonate, Mn(acac)3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Mn(C5H7O2)3
CAS Number:
Molecular Weight:
352.26
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

grade

technical

Assay

≥95% (KT)

mp

159-161 °C (dec.) (lit.)

SMILES string

CC(=O)\C=C(\C)O[Mn](O\C(C)=C/C(C)=O)O\C(C)=C/C(C)=O

InChI

1S/3C5H8O2.Mn/c3*1-4(6)3-5(2)7;/h3*3,6H,1-2H3;/q;;;+3/p-3/b3*4-3-;

InChI key

HYZQBNDRDQEWAN-LNTINUHCSA-K

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhan'ao Tan et al.
Physical chemistry chemical physics : PCCP, 14(42), 14589-14595 (2012-09-28)
A solution-processed vanadium oxide (s-VO(x)) anode buffer layer on an indium-tin-oxide (ITO) electrode was used instead of PEDOT:PSS for improving the stability and photovoltaic performance of the polymer solar cells (PSCs). The s-VO(x) layer was prepared by spin-coating a vanadyl
Hamid Golchoubian et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 85(1), 25-30 (2011-10-18)
Mixed-chelate copper(II) complexes with a general formula [Cu(acac)(diamine)]X where acac=acetylacetonate ion, diamine=N,N-dimethyl,N'-benzyl-1,2-diaminoethane and X=BPh(4)(-), PF(6)(-), ClO(4)(-) and BF(4)(-) have been prepared. The complexes were characterized on the basis of elemental analysis, molar conductance, UV-vis and IR spectroscopies. The complexes are
Zofia Kowalewska et al.
Talanta, 103, 66-74 (2012-12-04)
The aim of this work was to investigate the influence of magnesium acetylacetonate (MgA) on the signal of organic forms of vanadium in xylene solution by graphite furnace atomic absorption spectrometry. MgA alone or mixed with palladium acetylacetonate (PdA) was
Nathan A Mathew et al.
The journal of physical chemistry. A, 114(2), 817-832 (2009-12-03)
Nuclear magnetic resonance spectroscopy relies on using multiple excitation pulses to create multiple quantum coherences that provide great specificity for chemical measurements. Coherent multidimensional spectroscopy (CMDS) is the optical analogue of NMR. Current CMDS methods use three excitation pulses and
Channa R De Silva et al.
Journal of the American Chemical Society, 131(18), 6336-6337 (2009-04-17)
Nearly monodisperse lanthanide-doped magnetite nanoparticles were obtained by thermally decomposing a mixture of Fe(acac)(3) and Ln(acac)(3) (acac = acetylacetonate; Ln = Sm, Eu, Gd) in the presence of passivating surfactants. Magnetic studies revealed room-temperature ferromagnetic behaviors of these doped nanoparticles

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service