Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

07378

Sigma-Aldrich

Methyl 2-amino-4-chlorobenzoate

≥98.0% (HPLC/NT)

Synonym(s):

Methyl 4-chloroanthranilate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8ClNO2
CAS Number:
Molecular Weight:
185.61
Beilstein:
1072895
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98.0% (HPLC/NT)

form

powder or crystals

mp

66-71 °C

application(s)

peptide synthesis

SMILES string

COC(=O)c1ccc(Cl)cc1N

InChI

1S/C8H8ClNO2/c1-12-8(11)6-3-2-5(9)4-7(6)10/h2-4H,10H2,1H3

InChI key

YPSSCICDVDOEAI-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

07378-VAR:
07378-BULK:
07378-50G:
07378-10G:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service