Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

04934

Sigma-Aldrich

L-Serine benzyl ester benzenesulfonate (salt)

≥98.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13NO3 · C6H6O3S
CAS Number:
Molecular Weight:
353.39
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (HPLC)

form

powder or flakes

reaction suitability

reaction type: solution phase peptide synthesis

color

colorless to white

application(s)

peptide synthesis

SMILES string

OS(=O)(=O)c1ccccc1.N[C@@H](CO)C(=O)OCc2ccccc2

InChI

1S/C10H13NO3.C6H6O3S/c11-9(6-12)10(13)14-7-8-4-2-1-3-5-8;7-10(8,9)6-4-2-1-3-5-6/h1-5,9,12H,6-7,11H2;1-5H,(H,7,8,9)/t9-;/m0./s1

InChI key

AJKFQGHWMCPNTI-FVGYRXGTSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

04934-25G-F:
04934-5G-F:
04934-BULK-F:
04934-VAR-F:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service