03773
Ethylene glycol polymer-bound
extent of labeling: ~2.8 mmol/g diol loading
Synonym(s):
3-Benzylthio-1,2-propanediol polymer-bound, Polymer-supported diol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
PubChem Substance ID:
Recommended Products
extent of labeling
~2.8 mmol/g diol loading
matrix
crosslinked with 2% DVB
particle size
200-400 mesh
Other Notes
Polymer supported protecting-group reagent for aldehydes and ketones
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
P. Hodge et al.
Journal of the Chemical Society. Perkin Transactions 1, 2319-2319 (1983)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service