02730595
Larixyl acetate
primary reference standard
Synonym(s):
(αS,1S,4S,4aS,8aR)-4-(Acetyloxy)-α-ethenyldecahydro-α,5,5,8a-tetramethyl-2-methylene-1-naphthalenepropanol, Larixol acetate
About This Item
Recommended Products
grade
primary reference standard
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
HWI
application(s)
food and beverages
storage temp.
−20°C
SMILES string
CC(=O)O[C@H]1CC(=C)[C@H](CC[C@](C)(O)C=C)[C@@]2(C)CCCC(C)(C)[C@H]12
InChI
1S/C22H36O3/c1-8-21(6,25-16(3)23)13-10-17-15(2)14-18(24)19-20(4,5)11-9-12-22(17,19)7/h8,17-19,24H,1-2,9-14H2,3-7H3/t17-,18-,19-,21+,22+/m0/s1
InChI key
BSXKTJYNJOKMRN-TZWCSUAMSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Exact content by quantitative NMR can be found on the certificate.
Application
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
JAN Code
02730595-50MG:
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service