Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

35-0290

Sigma-Aldrich

Xylenol Orange

JIS special grade

Synonym(s):

3,3-Bis[N,N-bis(carboxymethyl)aminomethyl]-o-cresolsulfonephthalein

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H30N2O13SNa2
Molecular Weight:
716.62
EC Number:
UNSPSC Code:
41100000

grade

JIS special grade

form

solid

availability

available only in Japan

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

35-0290-0-100MG-J:
35-0290-1-1G-J:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sajitha Biju et al.
Plant physiology and biochemistry : PPB, 119, 250-264 (2017-09-17)
Silicon (Si) has been widely reported to have beneficial effect on mitigating drought stress in plants. However, the effect of Si on seed germination under drought conditions is still poorly understood. This research was carried out to ascertain the role
N G Martin Palmqvist et al.
Nanoscale research letters, 12(1), 631-631 (2017-12-21)
Yttrium doping-stabilized γ-Fe
Luisa Mota da Silva et al.
Inflammopharmacology, 24(6), 403-409 (2016-11-01)
This study investigated the effects of Rubus imperialis, a berry known as "amora-branca", in colitis dextran sulfate sodium (DSS)-induced in mice. Animals were treated orally with vehicle (water), 5-aminosalicylic acid (100 mg/kg) or methanolic extract from leaves of R. imperialis (MERI, 100 mg/kg)
Min Jung Kong et al.
Cell death & disease, 9(5), 488-488 (2018-04-27)
Mitochondrial NADP+-dependent isocitrate dehydrogenase (IDH2) plays an important role in the formation of NADPH, which is critical for the maintenance of mitochondrial redox balance. Cis-diamminedichloroplatinum II (cisplatin), an effective anticancer drug, induces oxidative stress-related nephrotoxicity, limiting its use. Therefore, we
Mohammad Wehbi et al.
Polymers, 12(9) (2020-09-03)
Novel triethoxysilane and dimethyl phosphonate functional vinylidene fluoride (VDF)-containing terpolymers, for potential applications in Eu ion extraction from water, were produced by conventional radical terpolymerization of VDF with vinyltriethoxylsilane (VTEOS) and vinyldimethylphosphonate (VDMP). Although initial attempts for the copolymerization of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service