Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

28-5860

Sigma-Aldrich

Sulfur

SAJ first grade, ≥98.0%

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
¥1,360

¥1,360


Available to ship onApril 07, 2025Details


Request a Bulk Order

Select a Size

Change View
500 G
¥1,360

About This Item

Empirical Formula (Hill Notation):
S
CAS Number:
Molecular Weight:
32.07
MDL number:
UNSPSC Code:
12141912
PubChem Substance ID:
grade:
SAJ first grade
form:
solid

¥1,360


Available to ship onApril 07, 2025Details


Request a Bulk Order

grade

SAJ first grade

vapor density

8.9 (vs air)

vapor pressure

1 mmHg ( 183.8 °C)
10 mmHg ( 246 °C)

Assay

≥98.0%

form

solid

autoignition temp.

450 °F

availability

available only in Japan

resistivity

2E23 μΩ-cm, 20°C

bp

444.7 °C (lit.)

mp

112.8 °C (rhombic) (lit.)
117-120 °C (lit.)
119.0 °C (monoclinic) (lit.)

storage temp.

15-25°C

SMILES string

[S]

InChI

1S/S

InChI key

NINIDFKCEFEMDL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 2: Flammable solids
Sulfur
Hazardous rank II

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

28-5860-5-500G-J:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Farhad Forouhar et al.
Nature chemical biology, 9(5), 333-338 (2013-04-02)
How living organisms create carbon-sulfur bonds during the biosynthesis of critical sulfur-containing compounds is still poorly understood. The methylthiotransferases MiaB and RimO catalyze sulfur insertion into tRNAs and ribosomal protein S12, respectively. Both belong to a subgroup of radical-S-adenosylmethionine (radical-SAM)
William Tam et al.
Journal of molecular biology, 425(14), 2450-2462 (2013-04-02)
The assembly of long non-contractile phage tails begins with the formation of the tail tip complex (TTC). TTCs are multi-functional protein structures that mediate host cell adsorption and genome injection. The TTC of phage λ is assembled from multiple copies
Chemoselective peptide ligation-desulfurization at aspartate.
Robert E Thompson et al.
Angewandte Chemie (International ed. in English), 52(37), 9723-9727 (2013-07-31)
Yu Kobayashi et al.
ACS nano, 9(4), 4056-4063 (2015-03-27)
Atomic-layer transition metal dichalcogenides (TMDCs) have attracted appreciable interest due to their tunable band gap, spin-valley physics, and potential device applications. However, the quality of TMDC samples available still poses serious problems, such as inhomogeneous lattice strain, charge doping, and
Gulluzar Bastug et al.
The Journal of organic chemistry, 78(18), 9303-9308 (2013-08-14)
The newly prepared complex [Pd(IPr*(OMe))(cin)(Cl)] provides high catalytic activity for carbon-sulfur cross-coupling reactions. Nonactivated and deactivated aryl halides were successfully coupled with a large variety of aryl- and alkylthiols using this well-defined palladium N-heterocyclic carbene (NHC) complex.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service