Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

28-3790

Sigma-Aldrich

Sodium phosphate monobasic dihydrate

JIS special grade, 99.0-102.0%

Synonym(s):

Sodium dihydrogen phosphate dihydrate

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
¥2,090

¥2,090


Available to ship onApril 07, 2025Details


Request a Bulk Order

Select a Size

Change View
500 G
¥2,090

About This Item

Linear Formula:
NaH2PO4 · 2H2O
CAS Number:
Molecular Weight:
156.01
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
grade:
JIS special grade
form:
solid

¥2,090


Available to ship onApril 07, 2025Details


Request a Bulk Order

grade

JIS special grade

Assay

99.0-102.0%

form

solid

availability

available only in Japan

pH

4.0-4.5 (25 °C, 50 g/L)

SMILES string

O.O.[Na+].OP(O)([O-])=O

InChI

1S/Na.H3O4P.2H2O/c;1-5(2,3)4;;/h;(H3,1,2,3,4);2*1H2/q+1;;;/p-1

InChI key

VBJGJHBYWREJQD-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

28-3790-5-500G-J:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Escuder-Gilabert et al.
Journal of chromatography. A, 1363, 331-337 (2014-07-22)
The design of experiments (DOE) is a good option for rationally limiting the number of experiments required to achieve the enantioresolution (Rs) of a chiral compound in capillary electrophoresis. In some cases, the modeled Rs after DOE analysis can be
Wenhu Zhou et al.
Analytical chemistry, 87(7), 4001-4007 (2015-03-11)
RNA-cleaving DNAzymes provide a unique platform for developing biosensors. However, a majority of the work has been performed in clean buffer solutions, while the activity of some important DNAzymes in biological sample matrices is still under debate. Two RNA-cleaving DNAzymes
Jan Muendges et al.
Biotechnology progress, 31(4), 925-936 (2015-04-11)
This article presents results of continuous multistage aqueous two-phase extraction of an immunoglobulin G1 from cell supernatant in a mixer-settler unit. An aqueous two-phase system consisting of polyethylene glycol 2000, phosphate salt, and water was applied without and with sodium
Soon Sik Kwon et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 92, 146-154 (2015-03-11)
We investigated the physicochemical properties of pH-sensitive hydroxyethyl cellulose (HEC)/hyaluronic acid (HA) complex hydrogels containing isoliquiritigenin (ILTG), and discussed potential applications as transdermal delivery systems for the treatment of skin lesions caused by pH imbalance. HA has skin compatibility and
Yoshiki Seto et al.
The Journal of pharmacology and experimental therapeutics, 354(2), 195-202 (2015-05-29)
This study aimed to qualify photosafety screening on the basis of photochemical and pharmacokinetic (PK) data on dermally applied chemicals. Six benzophenone derivatives (BZPs) were selected as model compounds, and in vitro photochemical/phototoxic characterization and dermal cassette-dosing PK study were

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service