Skip to Content
Merck
All Photos(1)

Documents

Safety Information

28-3630

Sigma-Aldrich

Sodium perborate tetrahydrate

SAJ first grade, ≥95.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaBO3 · 4H2O
CAS Number:
Molecular Weight:
153.86
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

SAJ first grade

Assay

≥95.0%

form

crystalline

availability

available only in Japan

pH

10.0-10.4 (25 °C, 10 g/L)

mp

60 °C (dec.) (lit.)

SMILES string

O.O.O.O.[Na+].[O-]B1OO1

InChI

1S/BO3.Na.4H2O/c2-1-3-4-1;;;;;/h;;4*1H2/q-1;+1;;;;

InChI key

IBDSNZLUHYKHQP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

PRTR

Class I Designated Chemical Substances

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

28-3630-5-500G-J:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Ghousia et al.
The Journal of clinical pediatric dentistry, 34(4), 303-308 (2010-09-14)
The objectives of this in vitro study, is to evaluate the influence of various dentin treatment procedures prior to bleaching namely, demineralization and demineralization in conjunction with deproteinization on the dentin permeability and bleaching efficacy. The study used a total
José A Murillo Pulgarín et al.
Phytochemical analysis : PCA, 22(1), 80-86 (2010-09-08)
The cobalt is an essential element for leguminous plants but may be harmful for other species; for that reason determination of Co(II) is very important for the management of polluted areas and for discover plants with capacity for the hyperaccumulation
D M P Spolidorio et al.
International journal of dental hygiene, 9(4), 279-283 (2011-03-02)
The aim of this study was to investigate the effectiveness of two alternatives methods for the disinfection of oral cleaning devices. One type of toothbrush and two types of tongue scrapers (steel and plastic) were tested in this study. Sixteen
In vitro evaluation of the efficacy of 4 denture cleanser tabs against oral bacteria.
D L Alves et al.
Bulletin du Groupement international pour la recherche scientifique en stomatologie & odontologie, 50(1), 31-35 (2011-01-01)
Ana Lucia Machado et al.
The Journal of prosthetic dentistry, 102(2), 115-122 (2009-08-01)
Microwave irradiation and immersion in chemical solutions have been recommended for denture disinfection. However, the effect of these procedures on the surface characteristics of denture base and reline resins has not been completely evaluated. The purpose of this study was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service