Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

15-1170

Sigma-Aldrich

Iron(II) chloride tetrahydrate

JIS special grade, 99.0-102.0%

Synonym(s):

Ferrous chloride tetrahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FeCl2 · 4H2O
CAS Number:
Molecular Weight:
198.81
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
JIS special grade
form:
fine crystals

grade

JIS special grade

vapor pressure

10 mmHg ( 693 °C)

Assay

99.0-102.0%

form

fine crystals

reaction suitability

reagent type: catalyst
core: iron

availability

available only in Japan

pH

2.5 (20 °C, 100 g/L)

SMILES string

O.O.O.O.Cl[Fe]Cl

InChI

1S/2ClH.Fe.4H2O/h2*1H;;4*1H2/q;;+2;;;;/p-2

InChI key

WSSMOXHYUFMBLS-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

15-1170-5-500G-J:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qinqin Xia et al.
The Journal of organic chemistry, 77(20), 9366-9373 (2012-10-03)
Iron-catalyzed direct C-N bond formation between azoles and amides is described. The oxidative coupling reactions of sp(3) C-H bonds adjacent to a nitrogen atom in amides and sulfonamides with the N-H bond in azoles proceeded smoothly in the presence of
Jiangfei Meng et al.
Journal of food science, 77(1), C8-14 (2011-12-21)
Wines made from 3 spine grape (Vitis davidii Foex) genotypes-Junzi 1# (JZ 1#), Junzi 2# (JZ 2#), and Liantang (LT)-and Cherokee rose (Rosa laevigata Michx., CR) were evaluated for their phenolics composition and antioxidant activities by several assays, including 2,2'-azino-bis-(3-ethylbenzothiazoline-6-sulfonic
Sofia Tresintsi et al.
Water research, 46(16), 5255-5267 (2012-07-25)
Various iron oxy-hydroxides were synthesized in a continuous flow kilogram-scale production reactor through the precipitation of FeSO(4) and FeCl(2) in the pH range 3-12 under intense oxidative conditions to serve as arsenic adsorbents. The selection of the optimum adsorbent and
Ju-Suk An et al.
Environmental technology, 35(13-16), 1668-1675 (2014-06-25)
The precipitation reaction between the orthophosphate and Fe2+ ions was studied to describe the optimum condition for the removal of orthophosphate from the aqueous solution. The effects of pH, Fe:P molar ratio, and alkalinity were evaluated for the initial orthophosphate
Angéla Hajdú et al.
Colloids and surfaces. B, Biointerfaces, 94, 242-249 (2012-03-01)
Magnetite nanoparticles (MNPs) were prepared by alkaline hydrolysis of Fe(II) and Fe(III) chlorides. Adsorption of polyacrylic acid (PAA) on MNPs was measured at pH=6.5±0.3 and I=0.01 M (NaCl) to find the optimal PAA amount for MNP stabilization under physiological conditions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service