Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

03-2680

Sigma-Aldrich

Bismuth standard solution

1 mg/mL Bi, suitable for atomic absorption spectrometry

Synonym(s):

Bismuth(III) nitrate solution

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
¥2,460

¥2,460


Available to ship onApril 01, 2025Details


Request a Bulk Order

Select a Size

Change View
100 ML
¥2,460

About This Item

MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

¥2,460


Available to ship onApril 01, 2025Details


Request a Bulk Order

form

liquid

availability

available only in Japan

concentration

1 mg/mL Bi

suitability

suitable for atomic absorption spectrometry

storage temp.

15-25°C

SMILES string

[Bi]

InChI

1S/Bi

InChI key

JCXGWMGPZLAOME-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

03-2680-6-100ML-J:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service