11934333001
Roche
mRNA Isolation Kit for Blood/Bone Marrow
sufficient for 100 isolation(s) (5 mL), kit of for up to 30 (100) isolations from 5 mL (1.5 mL) sample volumes, suitable for RT-PCR, suitable for Northern blotting, suitable for ELISA
Synonym(s):
mRNA
About This Item
Recommended Products
usage
sufficient for 100 isolation(s) (5 mL)
manufacturer/tradename
Roche
packaging
kit of for up to 30 (100) isolations from 5 mL (1.5 mL) sample volumes
technique(s)
ELISA: suitable
Northern blotting: suitable
RT-PCR: suitable
General description
- The mRNA Isolation Kit for Blood/Bone Marrow increases the chance of finding transcripts from rare cells:
- It reproducibly yields large quantities of mRNA (variance in yield is less than 10%).
- It can concentrate the isolated mRNA up to 250-fold (from 5 mL of whole blood to a single 20 μL sample, which may be directly amplified in a single RT-PCR).
- It does notrequire an initial cell-separation step, thus minimizing the potential for loss of single or rare target cells.
- The mRNA Isolation Kit for Blood/Bone Marrow is intended for use with a companion reagent, the RNA/DNA Stabilization Reagent for Blood/Bone Marrow (sold separately), which stabilizes and lyses biological samples. This reagent immediately inactivates nucleases, as well as stabilizes cell lysates in the field for transport back to the laboratory. It also simplifies the study of rare cells by:
- Immediately lysing the cells, thereby "suspending" the metabolic state of target cells and the level of expressed mRNA in them. This shields the cellular nucleic acids from changes that might occur when cells are removed from their natural environment.
- Protecting mRNA (and DNA) from nucleases immediately after sample collection.
Application
- RT-PCR
- Northern blotting
- Northern ELISA
- Ribonuclease protection assays
- cDNA library preparation
The intermediate nucleic acid fraction also can be processed by analytical techniques such as PCR. The mRNA Isolation Kit for Blood/Bone Marrow is particularly useful for the detection of rare cells like disseminated tumor cells by RT-PCR.
Features and Benefits
Isolate nucleic acids without prior cell separation, ensuring that mRNAs from all cells (even very rare cells) are represented in the final mRNA isolate.
Increase the efficiency of RT-PCR assays by removing PCR inhibitors (rRNA, tRNA, hemoglobin) from the mRNA template.
Obtain high yields of purified, intact mRNA, suitable for direct use in many research applications.
Increase the reliability and reproducibility of mRNA purification.
Easily stabilize blood or bone marrow using the RNA/DNA Stabilization Reagent for Blood/ Bone Marrow. Store samples for 12 months at -15 to -25°C or for one day at +2 to +8°C.
Components
- Magnetic Glass Particles (MGPs)
- MGP Wash Buffer
- MGP Elution Buffer
- Hybridization Buffer
- Oligo (dT)20 probe, biotin labeled
- Streptavidin Magnetic Particles (SMPs)
- SMP Washing Buffer
- Double-distilled Water
Quality
Note: 5 pg of MelJu cell mRNA is equivalent to about five melanoma cells.
Preparation Note
- The kit relies on magnetic glass particles and streptavidin-coated magnetic particles to isolate high-quality mRNA from stabilized peripheral blood and bone marrow aspirates.
- Lyse red blood cells and stabilize total nucleic acids with RNA/DNA Stabilization Reagent for Blood/Bone Marrow (separate reagent, not contained in the kit).
- Bind total nucleic acids to Magnetic Glass Particles and collect by centrifugation.
- Wash the Magnetic Glass Particles four times and collect them with a Magnetic Particle Separator after each wash.
- Elute total nucleic acids from the Magnetic Glass Particles. (Eluate may be used for DNA analysis.)
- Immobilize mRNA with biotinylated oligo(dT) and Streptavidin Magnetic Particles.
- Collect Streptavidin Magnetic Particles with a Magnetic Particle Separator and wash three times (as in step 3).
- Elute mRNA from Streptavidin-coated Magnetic Particles.
Other Notes
Lanes A : Sample stored at +2 to +8°C
Lanes B : Sample stored at +15 to +25°C
Lanes C : Sample stored at –15 to -25°C
Result: The same band was clearly visible in each sample. However, the sample stored at –15 to -25°C (lanes C) contained more beta-actin RNA than the sample stored at +2 to +8°C (lanes A), which in turn contained more than the sample stored at +15 to +25°C (lanes B).
A: bone marrow
B: blood
Result: Comparable results were obtained with both blood and bone marrow. The single sharp band indicates that the isolated RNA was not degraded.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
does not flash
Flash Point(C)
does not flash
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service