Skip to Content
Merck
All Photos(1)

Documents

Safety Information

SHF180UB25

Millipore

Hi-Flow Plus Membrane 180

width 25 mm , unbacked, pack of 1 slit roll

Synonym(s):

HF, Hi-Flow, Hi-Flow Plus HF180UB, immunoassay membrane, lateral flow membrane, nitrocellulose membrane, unbacked membrane

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NA.28

packaging

pack of 1 slit roll

Quality Level

manufacturer/tradename

Hi-Flow Plus

parameter

180 sec/4 cm capillary flow rate

roll L

100 m (328 ft)

width

25 mm , unbacked

Related Categories

General description

Hi-Flow Plus membranes are designed and optimised for use in rapid diagnostic lateral flow tests that require superior consistency and performance. Lateral flow tests faciliate quick and convenient detection / quantification of analytes such as antibodies and antigens present in biological samples, including blood, saliva, and urine. Hi-Flow™ Plus membranes are ideally suited for such applications. Available in a variety of capillary flow times and backing options, choosing Hi-Flow™ Plus membranes can improve assay performance and offer flexibility to meet a wide range of assay design requirements. Hi-Flow™ Plus 180 (HF180) is is suitable for tests where sensitivity is critical, the analyte of interest may present in limited quantities, and speed is not the primary concern. This Hi-Flow™ Plus 180 pack contains one 25mm x 100m slit roll of unbacked nitrocellulose membrane with a nominal capillary flow time of 180 seconds/4 cm.

Application

  • Hi-Flow Plus membrane is a critical raw material suitable for use in a broad range of Lateral Flow test applications, including Infectious Disease testing (COVID-19, HIV, Influenza, Malaria, STDs etc.), as well as in women’s health, biomarker detection, drugs of abuse testing, drug development, environmental testing, agricultural applications, food safety, product quality testing and animal health.
  • Hi-Flow Plus membranes are suitable for use with all lateral flow test techniques including competitive, sandwich, and multiplex detection assays.
  • They are optimised for use in both qualitative and quantitative test types and pair well with lateral flow detector particles including colloidal gold nanoparticles, dyed/ fluorescent microspheres etc.
  • Hi-Flow Plus membranes are suitable for use in lateral flow tests designed to detect analytes present in a broad range of samples matrices including, blood, saliva, urine, nasopharyngeal swabs, sweat and others.

Features and Benefits

FEATURES:
  • Consistent Flow & Surface Chemistry, Robust manufacturing process
  • Proprietary Surface Chemistry Optimised for Lateral Flow Applications
  • High Quality Level: MQ400
  • Trusted Partner and Brand with Decades of membrane manufacturing experience
  • High production capacity, high stock levels, multiple manufacturing lines
  • Convenient samples packs available in several formats
  • Manufacturing scale quantities available to purchase online with immediate shipping.
We offer a complete solution; along with Hi-Flow Plus membranes we offer raw materials across the entire lateral flow workflow as well as assay development services (include link to Lateral Flow Assay Development Services (sigmaaldrich.com))BENEFITS:
  • Consistent capillary flow time and membrane thickness
  • Superior surface morphology allows for consistent and even flow distribution of reagents across the membrane surface
  • Sharp, intense, and uniform test and control line formation
  • Excellent intra-lot and inter-lot consistency.
  • Better Assay sensitivity
  • Excellent reproducibility
  • Improved lateral flow test manufacturing yields.
  • Quick Delivery and Lead time.
  • No supply disruption
  • Excellent Technical support

Legal Information

Hi-Flow is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 5: Self-reactive substances
Nitric esters
Hazardous rank I
1st self-reactive materials

JAN Code

SHF180UB25:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service