SCT051
BioTracker Carbon Monoxide Probe 1 Live Cell Dye
Live cell imaging dye for cellular carbon monoxide (CO), a cell signalling molecule implicated in vasodilation, neurotransmission, anti-inflammatory and anti-apoptotic processes.
About This Item
Recommended Products
form
lyophilized
detection method
fluorometric
General description
Spectral Properties:Excitation: 450- 480nm
Emission: 525 nm (Green)
Research Category:Live Cell Dye
Materials Required but Not Supplied:
- PdCl2 (MilliporeSigma: Cat# 205885-1G or 520659-1G)
- CROM-3 (MilliporeSigma Cat# SML0496-10mg), a carbon monoxide-releasing molecule
Disclaimer
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
JAN Code
SCT051:
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service