RTTP14250
Polycarbonate Membrane Filter, 1.2 μm Pore Size
Isopore™, filter diam. 142 mm, hydrophilic
Synonym(s):
Isopore Membrane Filter
About This Item
Recommended Products
material
plain filter
polycarbonate membrane
white filter
sterility
non-sterile
feature
hydrophilic
manufacturer/tradename
Isopore™
Millipore
parameter
0.024 L/min-cm2-psi air flow rate
140 °C max. temp.
687 mL/min-cm2-psi water flow rate (maximum flow rate per 1.0 psi)
~175 mL/min-cm2 water flow rate at 10 psi (typical results)
diam.
142 mm
filter diam.
142 mm
thickness
24 μm
gravimetric extractables
<1%
refractive index
n/D 1.6
matrix
Isopore™
pore size
1.2 μm pore size
14.7 % porosity
bubble point
≥1.0 bar, air with water at 23 °C
shipped in
ambient
Looking for similar products? Visit Product Comparison Guide
General description
Features & Benefits:
•Membrane structure retains particles on the surface, simplifying counting and analysis
•Isopore membranes do not stain, resulting in low background interference
•Non-hygroscopic, allowing for rapid drying and reduced sample analysis time
•Translucent material does not require clearing for transmitted light microscopy; also available in brown variety
Applications:
Air Monitoring, Epifluorescent Microscopy, Chemotaxis Assays
Application
- Chemotaxis
- Bioassays
- Cytology
- Air monitoring
- Microplastics analysis
Caution
Legal Information
Storage Class Code
10-13 - German Storage Class 10 to 13
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service