Skip to Content
Merck
All Photos(8)

Documents

RDYTBCON01

Millipore

Pair of Female and Male Tube Quick Connectors

Adapter for bottle – dilutor quick connection. To be used in ReadyStream® workflow.

Synonym(s):

Tube Connector, Tube adaptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
13102028
NACRES:
NA.85

material

polysulfone (PSU)

Sterilization Compatibility

autoclavable compatible

packaging

pkg of 1 pair Of connectors

application(s)

food and beverages

General description

The autoclavable ReadyStream® tube connectors allow quick connection and disconnection of two tubes connected to a dilutor and a standard GL45 lab bottle serving as the media reservoir. In that way, the bottle can be filled by the ReadyStream® System at another place than where the dilutor is positioned to allow more flexibility. Then, after quick disconnecting the ReadyStream® System and transporting the filled bottle within the lab to the dispensing place, the bottle can be quick connected to the dilutor. Moreover, this setup allows filling a second GL45 lab bottle while the first bottle is connected to a dilutor, to parallelize the activities and to improve workflow efficiency.
For clean operations, a cap is contained in this reference as well, so that the connector can be sealed if needed.

Legal Information

READYSTREAM is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service