Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

QIA117

Sigma-Aldrich

Senescence Detection Kit

Synonym(s):

B-galactosidase senescence detection

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
Pricing and Inventory is not available. Please check the Merck website for more information.

usage

sufficient for 250 tests

Quality Level

packaging

pkg of 1 12-well plate(s)

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

input

sample type intact cells

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

General description

Senescence is thought to be a tumor suppressive mechanism and an underlying cause of aging. It represents an arrested state in which the cells remain viable, but not stimulated to divide by serum or by passage in culture. Senescent cells display an increase in cell size, pH-dependent expression of β-galactosidase (β-Gal) activity, and altered patterns of gene expression.
Useful for the histochemical detection of senescent cells. The failure of cells to become senescent may lead to tumor progression. Higher levels of β-Galactosidase is considered as a good marker for senescent cells. The mechanism of this is believed to be due to the increased lysosomal content of senescent cells, resulting in an increase of the lysosomal enzyme β-galactosidase. Cells can be observed using microscope.

Components

Fixative Solution, X-Gal, Staining Solution, Staining Supplement, and a user protocol.

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

Specifications

Assay Time: 30 min. plus overnight incubation

Principle

The Calbiochem® Senescence Detection Kit is useful for the histochemical detection of senescent cells. The failure of cells to become senescent may lead to tumor progression. The mechanism of this is believed to be the increased lysosomal content of senescent cells resulting in an increase of the lysosomal enzyme β-galactosidase.

Preparation Note

• PBS: Prepare a 1X PBS solution (not provided). Use at least 3 ml per well.• X-gal: Dissolve 20 mg X-gal in 1 ml of DMF (N-N-dimethylformamide, not provided) to prepare a 20X stock solution. Aliquot and freeze (-20°) the unused X-gal solution. This stock solution is stable for up to 1 month at -20°C (protect from light). Always store X-gal in a polypropylene or glass container. Do not use polystyrene.• Staining Solution Mix: While cells are incubating in Fixative Solution (step 2 below), prepare Staining Solution Mix in a polypropylene plastic tube by adding 470 µl of Staining Solution, 5 µl Staining Supplement, and 25 µl of 20 mg/ml X-gal in DMF.

Storage and Stability

Upon arrival store the entire contents of the kit at -20°C.

Other Notes

Kurz, D.J., et al. 2000. J. Cell Sci.113, 3613.
Dimri, G.P., et al. 1995. Proc. Natl. Acad. Sci. USA92, 9363.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 1B - Eye Dam. 1 - Muta. 2 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

PDSCL

Please refer to KIT Component information

PRTR

Please refer to KIT Component information

FSL

Please refer to KIT Component information

ISHL Indicated Name

Please refer to KIT Component information

ISHL Notified Names

Please refer to KIT Component information

Cartagena Act

Please refer to KIT Component information

JAN Code

キットコンポーネントの情報を参照してください


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ewelina Stelcer et al.
PloS one, 13(10), e0205691-e0205691 (2018-10-24)
Data on the response of chondrocytes differentiated from hiPSCs (hiPSC-DCHs) to ionizing radiation (IR) are lacking. The aim of present study was to assess DNA damage response (DDR) mechanisms of IR-treated hiPSC-DCHs. The following IR-response characteristics in irradiated hiPSC-DCHs were
Deana M Apple et al.
Aging, 11(1), 115-126 (2019-01-10)
The brain can generate new neurons from neural stem cells throughout life. However, the capacity for neurogenesis declines with age, reducing the potential for learning and repair. We explored the effects of calorie restriction, an established anti-aging intervention, on neural
Gabriela Ciapetti et al.
European spine journal : official publication of the European Spine Society, the European Spinal Deformity Society, and the European Section of the Cervical Spine Research Society, 21 Suppl 1, S10-S19 (2012-03-08)
Disc degeneration, and associated low back pain, are a primary cause of disability. Disc degeneration is characterized by dysfunctional cells and loss of proteoglycans: since intervertebral tissue has a limited capacity to regenerate, this process is at present considered irreversible.
Mary McMahon et al.
eLife, 8 (2019-09-04)
Small nucleolar RNAs (snoRNAs) are a diverse group of non-coding RNAs that direct chemical modifications at specific residues on other RNA molecules, primarily on ribosomal RNA (rRNA). SnoRNAs are altered in several cancers; however, their role in cell homeostasis as
Zixi Wang et al.
Nature cancer, 1(9), 909-922 (2021-08-14)
SWI/SNF chromatin remodelers play critical roles in development and cancer. The causal links between SWI/SNF complex disassembly and carcinogenesis are obscured by redundancy between paralogous components. Canonical cBAF-specific paralogs ARID1A and ARID1B are synthetic lethal in some contexts, but simultaneous

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service