NA-2ML-RU
Anti-N (clone BO3), Blood Grouping Reagent for Research Use
BIOSCOT™ Anti-N from the clone BO3, Murine IgG, Blood Grouping Reagent for Research Use
About This Item
biological source
mouse
antibody form
unpurified
clone
BO3, monoclonal
form
liquid
packaging
pkg of 2 mL
manufacturer/tradename
BIOSCOT™
concentration
2-6 mg/mL protein (Total Protein Range)
technique(s)
agglutination assay: suitable
pH
7-9
isotype
IgG
shipped in
ambient
storage temp.
2-8°C
General description
Application
Legal Information
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
JAN Code
NA-2ML-RU:
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service