Skip to Content
Merck
All Photos(1)

Documents

Safety Information

MABX8291

Sigma-Aldrich

Anti-Hepatitis B Core Antigen (HBcAg) Antibody, (1mg)

clone 1, Chemicon®, from mouse

Synonym(s):

Hepatitis B core antigen

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

1, monoclonal

purified by

affinity chromatography

species reactivity

virus, hepatitis B virus (Core Ag)

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunodiffusion: suitable

isotype

IgG2a

General description

Hepatitis B core antigen is an indicator of active HBV viral replication. Detection of the HBcAg along with HBeAg is a suggested addition at times when/if the HBsAg might be undetectable.

Specificity

Core antigen of Hepatitis B virus (serotypes adw, ayw)

Immunogen

Recombinant full-length HBcAg

Application

Detect HBcAg using mouse monoclonal Anti-HBcAg, Cat No MABX8291-1KC, validated for use in ELISA and Laterla Flow Assay.
Research Category
Infectious Diseases

Quality

Detect HBcAg using mouse monoclonal Anti-HBcAg, Cat No MABX8291-1KC, validated for use in ELISA

Physical form

Affinity chromotography purification
Purified mouse monoclonal antibody in buffer containing 10 mM Tris-HCl, pH 8.0, 0.05% NaN3

Storage and Stability

2-8C. For long-term storage, freeze at -20C.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

MABX8291-1KC:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service