Skip to Content
Merck
All Photos(2)

Documents

Safety Information

MABT189

Sigma-Aldrich

Anti-Delta3 Antibody, clone 1E7.2

clone 1E7.2, from mouse

Synonym(s):

Delta-like protein 3, Drosophila Delta homolog 3, Delta3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1E7.2, monoclonal

species reactivity

human

technique(s)

flow cytometry: suitable
western blot: suitable

isotype

IgMκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... DLL3(10683)

Related Categories

General description

DLL3, also known as Delta like ligand 3, is a transmembrane Delta-like protein principally found in the Golgi apparatus and endosomes where it promotes Notch family receptor degradation to inhibit primary neurogenesis. DLL3 is one of five DSL proteins that bind to the Notch receptor. Notch signaling pathways play key roles in cell-fate determination and differentiation in many tissues during embryonic and postnatal development. Notch ligands family members are characterized by a DSL domain, EGF repeats, and a transmembrane domain. Mutations in the DLL3 gene causes autosomal recessive spondylocostal dysostosis.

Immunogen

Linear peptide corresponding to human Delta3.

Application

Flow Cytometry Analysis: 0.1 µg from a representative lot detected Delta3 in HEK293 cells.
Research Category
Cell Structure
Research Sub Category
ECM Proteins
This Anti-Delta3 Antibody, clone 1E7.2 is validated for use in WB, FC for the detection of Delta3.

Quality

Evaluated by Western Blot in human liver tissue lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected Delta3 on 10 µg of human liver tissue lysate.

Target description

~65 kDa observed

Physical form

Format: Purified
Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human liver tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

MABT189:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service