Skip to Content
Merck
All Photos(1)

Documents

Safety Information

MABN628

Sigma-Aldrich

Anti-Cox-1, clone 5C5.1 Antibody

clone 5C5.1, 1 mg/mL, from mouse

Synonym(s):

Cytochrome c oxidase subunit 1, Cytochrome c oxidase polypeptide I

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

5C5.1, monoclonal

species reactivity

human

concentration

1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MT-CO1(4512)

General description

Cox-1, also known as Cytochrome C oxidase subunit 1, Cytochrome C oxidase polypeptide I, and encoded by the gene MT-CO1, CO1, COX1, MTCO1, is the catalytic subunit of Cytochrome C oxidase that forms the final reduction of oxygen to water in the respiratory chain. Subunits 1-3 form the functional core. Electrons originating in Cytochrome C are transferred via the copper A center of subunit 2 and the heme A of subunit 1 to the bimetallic center formed by the heme A3 and Copper B. The Cox-1 gene is encoded in the mitochondrion and the protein localizes to the inner mitochondrion membrane. It is expressed in all cells that contain functional mitochondria and mutations in Cox-1 are associated with various mitochondrial dysfunctional diseases that manifest themselves in defective vision, anemia, cardiomyopathy, muscle weakness and even some forms of cancer.

Immunogen

KLH-conjugated linear peptide corresponding to human Cox-1.

Application

Detect Cox-1 using this Anti-Cox-1 antibody, clone 5C5.1 validated for use in western blotting & IHC.
Immunohistochemistry Analysis: A 1:500-2,000 dilution from a representative lot detected Cox-1 in human thalamus, human kidney and human cerebellum tissue.
Research Category
Neuroscience
Research Sub Category
Developmental Signaling

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected Cox-1 in 10 µg of HeLa cell lysate.

Target description

~65 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

MABN628:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service