Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

MABN2303

Sigma-Aldrich

Anti-GABARAP Antibody, clone 8H5

Synonym(s):

Gamma-aminobutyric acid receptor-associated protein;GABA(A) receptor-associated protein;MM46

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rat

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

8H5, monoclonal

mol wt

calculated mol wt 13.92 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100 μg

technique(s)

ELISA: suitable
dot blot: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
western blot: suitable

isotype

IgG2aκ

epitope sequence

N-terminal

Protein ID accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... GABARAP(11337)

General description

Gamma-aminobutyric acid receptor-associated protein (UniProt: O95166; also known as GABA(A) receptor-associated protein, MM46) is encoded by the GABARAP (also known as FLC3B, HT004) gene (Gene ID: 11337) in human. GABARAP, is a member of the highly conserved family of mammalian autophagy-related 8 protein (ATG8), is implicated in a variety of intracellular transport processes and in autophagy where it is involved in late stage autophagosome maturation. Like other ATG8s, GABARAP can also undergo lipidation by a ubiquitin-like conjugation system, which then promotes its association to autophagosomal membranes. The precursor form of human GABARAP is processed by ATG4 to expose the C-terminal glycine to generate the cytosolic form GABARAP-I, which is then activated by APG7L/ATG7, transferred to ATG3 and conjugated to phosphatidylethanolamine (PE) phospholipid to produce the membrane-bound GABARAP-II form. Under nutrient rich conditions, GABARAP is reported to accumulate in the pericentriolar material from which it can be translocated to forming autophagosomes during starvation. Its expressed is observed in heart, brain, placenta, liver, skeletal muscle, kidney, and pancreas. It is also reported to serve as a ubiquitin-like modifier that plays a role in intracellular transport of GABA(A) receptors. Clone 8H5 recognizes an epitope in the N-terminal region and exhibits high specificity for GABARAP without any cross-reactivity for GABARAPL1, -L2 or LC3. (Ref.: Simons, IM., et al. (2019). Sci. Rep. 9; 256; Kabeya, Y., et al. (2004). J. Cell Sci. 117(13); 2805-2812).

Specificity

Clone 8H5 is a rat monoclonal antibody that specifically detects GABA(A) receptor-associated protein. It targets an epitope within the N-terminal domain.

Immunogen

GST-tagged full-length human recombinant GABA(A) receptor-associated protein.

Application

Quality Control Testing

Evaluated by Immunocytochemistry in serum starved HEK293 cells treated with Bafilomycin A.

Immunocytochemistry Analysis (ICC): A 1:100 dilution of this antibody detected GABARAP in serum starved HEK293 cells treated with Bafilomycin A (100 nM, 4 h).

Tested Applications


Immunocytochemistry Analysis: A representative lot detected GABARAP in Immunocytochemistry applications (Simons, I.M., et al. (2019). Sci Rep. 9(1): 526).

Immunofluorescence Analysis: A representative lot detected GABARAP in Immunofluorescence applications (Simons, I.M., et al. (2019). Sci Rep. 9(1): 526).

ELISA Analysis: A representative lot detected GABARAP in ELISA applications (Simons, I.M., et al. (2019). Sci Rep. 9(1): 526).

Dot Blot: A representative lot detected GABARAP in Dot Blotting applications (Simons, I.M., et al. (2019). Sci Rep. 9(1): 526).

Western Blotting Analysis: A representative lot detected GABARAP in Western Blotting applications (Simons, I.M., et al. (2019). Sci Rep. 9(1): 526).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-GABARAP, clone 8H5, Cat. No. MABN2303, is a rat monoclonal antibody that detects GABARAP and is tested for use in DOT Blot, ELISA, Immunocytochemistry, Immunofluorescence, and Western Blotting.

Physical form

Purified rat monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

MABN2303-100UG:
MABN2303-25UG:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service