Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

MABF2795

Sigma-Aldrich

Anti-IL-9 Antibody, clone MH9A4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.74

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

MH9A4, monoclonal

mol wt

observed mol wt ~57 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100 μL

technique(s)

ELISA: suitable
flow cytometry: suitable

isotype

IgG2b

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

shipped in

2-8°C

target post-translational modification

unmodified

Gene Information

mouse ... IL9(3578)

General description

Interleukin-9 (UniProt: P15248; also known as IL-9, Cytokine P40, T-cell growth factor P40) is encoded by the IL9 gene (Gene ID: 3578) in human. IL-9 is a pleiotropic cytokine that plays a significant role in tolerance and inflammation. It is expressed by a variety of immune cells. It is synthesized with a signal peptide (aa 1-18), which is subsequently cleaved off to generate the mature form. It supports the growth of mast cells, T cells, and several other cell types, including erythroid progenitors, fetal thymocytes, human megakaryoblastic leukemic cells, and myeloid precursor cells. It is also reported to supports IL-2 independent and IL-4 independent growth of helper T-cells. It also plays a role in promoting IgE production by B cells. IL-9 actions are mediated via its binding to IL-9R that results in the formation the IL-9R heterocomplex. Following its binding to the IL-9R , the SHC is phosphorylated, which then binds to Grb2 that subsequently activates SoS, a GTP exchange factor for Ras GTPases. This results in the activation of Ras GTPase that activates Raf kinase followed by the activation of MEK1/2 and ERK1/2. Although IL-9 is reported to activate the MAPK pathway in several lymphoid and hematopoietic cells, this activation is weaker compared to other cytokines. Elevated levels of IL-9 and IL-9R are observed in lungs of asthmatic subjects. (Ref.: Chakraborty, S., et al. (2019). Int. J. Mol. Sci. 20(9); 2113).

Specificity

Clone MH9A4 is a mouse monoclonal antibody that detects human Interleukin-9.

Immunogen

Ovalbumin-conjugated recombinant human Interleukin-9.

Application

Quality Control Testing

Evaluated by ELISA with recombinant human IL-9.

ELISA Analysis:: Various dilution of this antibody detected recombinant human Interleukin-9 (IL-9).

Tested Applications

Flow Cytometry Analysis: A representative lot detected IL-9 in Flow Cytometry applications (Lozano, E., et al. (2012). J Immunol.;188(8):3869-75; Muls, N., et al. (2012). J Neuroimmunol.;243(1-2):73-80).

ELISA Analysis: A representative lot detected IL-9 in ELISA applications (Jenmalm, M.C., et al. (2001). Clin Exp Allergy.;31(10):1528-35; Faulkner, H., et al. (2002). J Infect Dis.;185(5):665-72; Bottcher, M.F., et al. (2003). Pediatr Allergy Immunol.;14(5):345-50; Holz, L.E., et al. (2005). J Inflamm (Lond).;2:3; Chang, H.C., et al. (2010). Nat Immunol.;11(6):527-34; Stephens, G.L., et al. (2011). Eur J Immunol.;41(4):952-62; Lozano, E., et al. (2012). J Immunol.;188(8):3869-75; Muls, N., et al. (2012). J Neuroimmunol.;243(1-2):73-80).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-IL-9, clone MH9A4, Cat. No. MABF2795, is a mouse monoclonal antibody that detects Interleukin-9 and is tested for use in ELISA and Flow Cytometry.

Physical form

Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommend storage at +2°C to +8°C. For long term storage antibodies can be kept at -20°C. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

MABF2795-100UL:
MABF2795-25UL:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service