Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

MABF2678

Sigma-Aldrich

Anti-KSHV gpK8.1A Antibody, clone 4A4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

4A4, monoclonal

mol wt

calculated mol wt 24.81 kDa
observed mol wt ~35 kDa

purified by

using protein G

species reactivity

virus

packaging

antibody small pack of 100 μL

technique(s)

ELISA: suitable
electron microscopy: suitable
immunocytochemistry: suitable
inhibition assay: suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

vaccinia virus ... K8.1(4961469)

General description

Kaposi′s sarcoma-associated herpesvirus glycoprotein K8.1 (UniProt: O36551; also known as Glycoprotein K8.1, Glycoprotein K8.1A, Glycoprotein gp35/37, K8.1, K8.1 protein) is encoded by the K8.1 (also known as HHV8GK18_gp55) gene (Gene ID: 4961469) in Kaposi′s sarcoma-associated herpesvirus (KSHV). Kaposi sarcoma virus is an oncogenic human g-herpesvirus that has been identified in AIDS patients in association with Kaposi′s sarcoma. It can persist latently in host cells while expressing only a limited set of viral genes and maintaining its genome as extrachromosomal plasmids. This virus encodes two immunogenic class I glycoproteins, K8.1A and K8.1B. These glycoproteins have several serine and threonine residues that serves as potential sites for O-glycosylation. The 228-amino-acid-long K8.1A open reading frame (ORF) contains four N-glycosylation sites and the 167-amino-acid-long K8.1B ORF contains three N-glycosylation sites. Glycoprotein K8.1A is reported to mediate KSHV binding to target cells via interaction with heparan sulfate. This protein is involved in binding to target cells but is not required for viral entry into cells. Clone 4A4 detects both K8.1A and K8.1B glycoprotein and is shown to react with infected cell membranes and virion envelope. This antibody is shown to partially inhibit KSHV infection in HEK293 cells. (Ref.: Faure, A., et al. (2019). Proc. Natl. Acad. Sci. USA. 116(33); 16519-16528; Zhu, L., et al. (1999). Virology 262(1), 237-249).

Specificity

Clone 4A4 is a mouse monoclonal antibody that detects Glycoprotein K8.1 of Kaposi′s sarcoma-associated herpesvirus.

Immunogen

Phorbol-12-myristate-13-acetate TPA))- induced BCBL-1 (Body Cavity Based Lymphoma-1) cells.

Application

Quality Control Testing

Evaluated by Western Blotting in Phorbol-12-myristate-13-acetate (TPA)-induced BCBL-1 cells.

Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected Kaposi′s sarcoma-associated herpesvirus (KSHV) in Phorbol-12-myristate-13-acetate (TPA)- induced BCBL-1 cells.

Tested Applications

Inhibition: A representative lot of this antibody inhibited infection by KSHV in HEK293 cells.(Faure, A., et al. (2019). Proc Natl Acad Sci USA. 116(33):16519-16528).

Electron Microscopy: A representative lot of this antibody detected K8.1A glycoprotein in TPA stimulated BCBL-1 cells. (Zhu, L., et al. (1999). Virology. 262(1):237-49).

Western Blotting Analysis: A representative lot detected KSHV gpK8.1A in Western Blotting applications (Zhu, L., et al. (1999). Virology. 262(1):237-49).

ELISA Analysis: A representative lot detected KSHV gpK8.1A in ELISA applications (Zhu, L., et al. (1999). Virology. 262(1):237-49).

Immunocytochemistry Analysis: A representative lot detected KSHV gpK8.1A in Immunocytochemistry applications (Zhu, L., et al. (1999). Virology. 262(1):237-49).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-KSHV gpK8.1A, clone 4A4, Cat. No. MABF2678, is a mouse monoclonal antibody that detects Kaposi′s sarcoma-associated herpesvirus (KSHV) and is tested in Electron Microscopy, ELISA, Immunocytochemistry, Inhibition Assay, and Western Blotting.

Physical form

Purified mouse monoclonal antibody IgG1 in PBS without azide.

Storage and Stability

Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

MABF2678-100UL:
MABF2678-25UL:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service