Skip to Content
Merck
All Photos(3)

Key Documents

Safety Information

MABE463M

Sigma-Aldrich

Anti-methylated TLS/FUS Antibody, clone 9G6

clone 9G6, from rat

Synonym(s):

RNA-binding protein FUS, 75 kDa DNA-pairing protein, Oncogene FUS, Oncogene TLS, POMp75, Translocated in liposarcoma protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

9G6, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

target post-translational modification

methylation (not specified)

Gene Information

human ... FUS(2521)

General description

RNA-binding protein FUS (UniProt: also known as 75 kDa DNA-pairing protein, Oncogene FUS, Oncogene TLS, POMp75, Translocated in liposarcoma protein) is encoded by the FUS (also known as TLS) gene (Gene ID: 2521) in human. Fused in sarcoma/translocated in liposarcoma FUS/TLS is a multi-functional nuclear protein that binds both single-stranded and double-stranded DNA and promotes ATP-independent annealing of complementary single-stranded DNAs and D-loop formation in super helical double-stranded DNA. It is involved in a variety of cellular functions including transcription, protein translation, RNA splicing, and transport and may play a role in maintenance of genomic integrity. A chromosomal aberration involving FUS is shown to cause of acute myeloid leukemia (AML). Mutations in FUS gene have been linked to Amyotrophic lateral sclerosis 6, with or without frontotemporal dementia that affects upper motor neurons in the brain and lower motor neurons in the brain stem and spinal cord, resulting in fatal paralysis. (Ref.: Lagier-Tourenne, C., et al. (2010). Hum. Mol. Genet. 19(1); R46-R64).

Specificity

Clone 9G6 detects methylated RNA-binding protein FUS in human cells. It targets an epitope with in 31 amino acids from the C-terminal region.

Immunogen

Ovalbumin-conjugated linear peptide corresponding to 31 amino acids from the C-terminal region corresponding to the RGG3 domain of FUS (with all arginines methylated).

Application

Anti-methylated TLS/FUS, clone 9G6, Cat. No. MABE463, is a highly specific rat monoclonal antibody that targets methylated RNA-binding protein FUS and has been tested in Western Blotting.
Research Category
Epigenetics & Nuclear Function
Western Blotting Analysis: 0.05 ug/mL frrom a representative lot detected methylated TLS/FUS in 10 µg of HeLa cell lysate.

Western Blotting Analysis: 0.05 ug/mL frrom a representative lot detected methylated TLS/FUS in 10 µg of lysate from HeLa cells pretreated with AdOx. (Dormann, D., et al. (2012). The EMBO J. 31(22):4258-75)

Western Blotting Analysis: A representative lot detected methylated TLS/FUS in Western Blotting applications (Suárez-Calvet, M., et. al. (2016). Acta Neuropathol. 131(4):587-604).


Quality

Evaluated by Western Blotting in HepG2 cell lysate.

Western Blotting Analysis: 0.05 ug/mL of this antibody detected methylated TLS/FUS in 10 µg of HepG2 cell lysate.

Target description

~70 kDa observed; 53.43 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified rat monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

MABE463-25UG:
MABE463:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service